4-Benzyloxazolidine-2,5-dione 95%
Lieferant: Apollo Scientific
OR953604-1G
APOSOR953604-1GEA
52.2
EUR
APOSOR953604-1G
4-Benzyloxazolidine-2,5-dione 95%
4-Benzyloxazolidine-2,5-dione
Formel:
C₁₀H₉NO₃ Molecular Weight: 191,183 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18375238 CAS-Nummer: 583-47-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...