5-Cyano-2,3-dihydro-1H-indole 98%

Lieferant: Apollo Scientific

Synonyme: Indoline-5-carbonitrile

OR953724-1G OR953724-250MG OR953724-5G
APOSOR953724-1GEA 154 EUR
APOSOR953724-1G APOSOR953724-250MG APOSOR953724-5G
5-Cyano-2,3-dihydro-1H-indole 98%
5-Cyano-2,3-dihydro-1H-indole
Formel: C₉H₈N₂
Molecular Weight: 144,173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07371626
CAS-Nummer: 15861-23-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR