Hexahydro-1H-azepin-4-ol hydrochloride 95%

Lieferant: Apollo Scientific

Synonyme: Azepan-4-ol hydrochloride

OR953743-1G OR953743-5G
APOSOR953743-1GEA 91.5 EUR
APOSOR953743-1G APOSOR953743-5G
Hexahydro-1H-azepin-4-ol hydrochloride 95%
Hexahydro-1H-azepin-4-ol hydrochloride
Formel: C₆H₁₃NO·ClH
Lagertemperatur: Kühlschrank
MDL: MFCD11053644
CAS-Nummer: 1159823-34-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR