Cyclopropyl(pyridin-3-yl)methanamine 95%

Lieferant: Apollo Scientific

Synonyme: 1-Cyclopropyl-1-(3-pyridyl)methylamine

OR953745-1G OR953745-250MG
APOSOR953745-1GEA 270 EUR
APOSOR953745-1G APOSOR953745-250MG
Cyclopropyl(pyridin-3-yl)methanamine 95%
Cyclopropyl(pyridin-3-yl)methanamine
Formel: C₉H₁₂N₂
Molecular Weight: 148,205 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD05191947
CAS-Nummer: 535925-69-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR