1-(3-Methoxyphenyl)ethylamine hydrochloride 95%
Lieferant: Apollo Scientific
Synonyme:
1-(3-Methoxyphenyl)ethylamine Hydrochloride
OR953788-25G
OR953788-5G
APOSOR953788-25GEA
520
EUR
APOSOR953788-25G
APOSOR953788-5G
1-(3-Methoxyphenyl)ethylamine hydrochloride 95%
1-(3-Methoxyphenyl)ethylamine hydrochloride
Formel:
C₉H₁₃NO·ClH Molecular Weight: 187.667 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12198854 CAS-Nummer: 854184-18-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...