Indole-7-carboxylic acid ethyl ester 95%
Lieferant: Apollo Scientific
Synonyme: Ethyl 1H-indole-7-carboxylate
OR953799-1G OR953799-5G
APOSOR953799-1GEA 178 EUR
APOSOR953799-1G APOSOR953799-5G
Indole-7-carboxylic acid ethyl ester 95%
Indole-7-carboxylic acid ethyl ester
Formel: C₁₁H₁₁NO₂
Molecular Weight: 189.211 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00238711
CAS-Nummer: 205873-58-1