4-Chloro-2-(dimethylamino)-1,3-thiazole-5-carbaldehyde 95%
Lieferant: Apollo Scientific
OR953894-1G
OR953894-5G
APOSOR953894-1GEA
168
EUR
APOSOR953894-1G
APOSOR953894-5G
4-Chloro-2-(dimethylamino)-1,3-thiazole-5-carbaldehyde 95%
4-Chloro-2-(dimethylamino)-1,3-thiazole-5-carbaldehyde
Formel:
C₆H₇ClN₂OS Molecular Weight: 190.651 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD07852062 CAS-Nummer: 129865-54-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...