4-[(Methylsulphanyl)methyl]benzoic acid 98%
Lieferant: Apollo Scientific
OR954025-1G
OR954025-250MG
APOSOR954025-1GEA
435
EUR
APOSOR954025-1G
APOSOR954025-250MG
4-[(Methylsulphanyl)methyl]benzoic acid 98%
4-[(Methylsulphanyl)methyl]benzoic acid
Formel:
C₉H₁₀O₂S Molecular Weight: 182,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06373463 CAS-Nummer: 67003-48-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...