6-Methyl-1H-indazol-5-ol 95%
Lieferant: Apollo Scientific
OR954145-1G
OR954145-250MG
APOSOR954145-1GEA
719
EUR
APOSOR954145-1G
APOSOR954145-250MG
6-Methyl-1H-indazol-5-ol 95%
6-Methyl-1H-indazol-5-ol
Formel:
C₈H₈N₂O Molecular Weight: 148.162 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD08704815 CAS-Nummer: 478832-60-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...