Methyl 5-hydroxy-pyrazole-3-carboxylate 96%

Lieferant: Apollo Scientific

Synonyme: Methyl 5-hydroxy-1H-pyrazole-3-carboxylate

OR954158-1G OR954158-5G
APOSOR954158-1GEA 68.3 EUR
APOSOR954158-1G APOSOR954158-5G
Methyl 5-hydroxy-pyrazole-3-carboxylate 96%
Methyl 5-hydroxy-pyrazole-3-carboxylate
Formel: C₅H₆N₂O₃
Molecular Weight: 142.113 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878841
CAS-Nummer: 1018446-60-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR