4-Bromo-1-methanesulphonyl-2-methylbenzene 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-methyl-1-(methylsulphonyl)benzene
OR954273-1G
OR954273-5G
APOSOR954273-1GEA
89.9
EUR
APOSOR954273-1G
APOSOR954273-5G
4-Bromo-1-methanesulphonyl-2-methylbenzene 96%
4-Bromo-1-methanesulphonyl-2-methylbenzene
Formel:
C₈H₉BrO₂S Molecular Weight: 249.125 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22549347 CAS-Nummer: 99768-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...