4-Bromo-2-methoxy-1-(methylsulphonyl)benzene 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-2-methoxyphenyl methyl sulphone
OR954279-1G
OR954279-5G
APOSOR954279-1GEA
216
EUR
APOSOR954279-1G
APOSOR954279-5G
4-Bromo-2-methoxy-1-(methylsulphonyl)benzene 98%
4-Bromo-2-methoxy-1-(methylsulphonyl)benzene
Formel:
C₈H₉BrO₃S Molecular Weight: 265,124 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22586698 CAS-Nummer: 1095544-87-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...