Ethyl benzimidazole-5-carboxylate 98%
Lieferant: Apollo Scientific
OR954312-1G
OR954312-5G
APOSOR954312-1GEA
183
EUR
APOSOR954312-1G
APOSOR954312-5G
Ethyl benzimidazole-5-carboxylate 98%
Ethyl benzimidazole-5-carboxylate
Formel:
C₁₀H₁₀N₂O₂ Molecular Weight: 190.199 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027455 CAS-Nummer: 58842-61-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...