2-Amino-5-ethoxybenzonitrile 98%

Lieferant: Apollo Scientific

OR954345-1G OR954345-5G
APOSOR954345-1GEA 270 EUR
APOSOR954345-1G APOSOR954345-5G
2-Amino-5-ethoxybenzonitrile 98%
2-Amino-5-ethoxybenzonitrile
Formel: C₉H₁₀N₂O
Molecular Weight: 162,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17168771
CAS-Nummer: 549488-78-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR