Ethyl 1H-1,2,3-benzotriazole-5-carboxylate 98%
Lieferant: Apollo Scientific
Synonyme:
Ethyl 1H-benzo[d][1,2,3]triazole-5-carboxylate
OR954350-1G
OR954350-5G
APOSOR954350-1GEA
155
EUR
APOSOR954350-1G
APOSOR954350-5G
Ethyl 1H-1,2,3-benzotriazole-5-carboxylate 98%
Ethyl 1H-1,2,3-benzotriazole-5-carboxylate
Formel:
C₉H₉N₃O₂ Molecular Weight: 191,187 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00963122 CAS-Nummer: 73605-91-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...