4,4-Dimethyl-1,3-dihydroquinolin-2-one 98%

Lieferant: Apollo Scientific

OR954434-10G OR954434-1G OR954434-5G
APOSOR954434-10GEA 647 EUR
APOSOR954434-10G APOSOR954434-1G APOSOR954434-5G
4,4-Dimethyl-1,3-dihydroquinolin-2-one 98%
4,4-Dimethyl-1,3-dihydroquinolin-2-one
Formel: C₁₁H₁₃NO
Molecular Weight: 175,227 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11040447
CAS-Nummer: 76693-04-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR