3-(1,3,4-Oxadiazol-2-yl)phenol 98%
Lieferant: Apollo Scientific
OR954528-1G
OR954528-5G
APOSOR954528-1GEA
190
EUR
APOSOR954528-1G
APOSOR954528-5G
3-(1,3,4-Oxadiazol-2-yl)phenol 98%
3-(1,3,4-Oxadiazol-2-yl)phenol
Formel:
C₈H₆N₂O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD06643238 CAS-Nummer: 5378-29-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...