5-(3-Hydroxyphenyl)-3H-1,3,4-oxadiazol-2-one 98%

Lieferant: Apollo Scientific

OR954529-1G OR954529-250MG
APOSOR954529-1GEA 754 EUR
APOSOR954529-1G APOSOR954529-250MG
5-(3-Hydroxyphenyl)-3H-1,3,4-oxadiazol-2-one 98%
5-(3-Hydroxyphenyl)-3H-1,3,4-oxadiazol-2-one
Formel: C₈H₆N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD19673427
CAS-Nummer: 79302-77-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR