2-(2-Bromophenyl)-5-(chloromethyl)-1,3,4-oxadiazole 98%

Lieferant: Apollo Scientific
Achtung

OR954563-1G OR954563-25G OR954563-5G
APOSOR954563-1GEA 106 EUR
APOSOR954563-1G APOSOR954563-25G APOSOR954563-5G
2-(2-Bromophenyl)-5-(chloromethyl)-1,3,4-oxadiazole 98%
2-(2-Bromophenyl)-5-(chloromethyl)-1,3,4-oxadiazole
Formel: C₉H₆BrClN₂O
Molecular Weight: 273,514 g/mol
Schmelzpunkt: 110…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03981004
CAS-Nummer: 733044-96-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR