1-tert-Butylpyrazole-4-carbaldehyde 96%
Lieferant: Apollo Scientific
Synonyme:
1-(tert-Butyl)-1H-pyrazole-4-carbaldehyde
OR954587-10G
OR954587-1G
OR954587-5G
APOSOR954587-10GEA
502
EUR
APOSOR954587-10G
APOSOR954587-1G
APOSOR954587-5G
1-tert-Butylpyrazole-4-carbaldehyde 96%
1-tert-Butylpyrazole-4-carbaldehyde
Formel:
C₈H₁₂N₂O Molecular Weight: 152,194 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09939615 CAS-Nummer: 127949-06-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...