trans-Cyclohexane-1,3-dicarboxylic acid 95%
Lieferant: Apollo Scientific
OR954683-1G
OR954683-5G
APOSOR954683-1GEA
53.8
EUR
APOSOR954683-1G
APOSOR954683-5G
trans-Cyclohexane-1,3-dicarboxylic acid 95%
trans-Cyclohexane-1,3-dicarboxylic acid
Formel:
C₈H₁₂O₄ Molecular Weight: 172,178 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01311241 CAS-Nummer: 2305-30-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...