4-(Thiophen-3-yl)phenol 97%
Lieferant: Apollo Scientific
OR954763-1G
OR954763-5G
APOSOR954763-1GEA
114
EUR
APOSOR954763-1G
APOSOR954763-5G
4-(Thiophen-3-yl)phenol 97%
4-(Thiophen-3-yl)phenol
Formel:
C₁₀H₈OS Molecular Weight: 176,235 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06798133 CAS-Nummer: 29886-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...