4-(2-Methylphenyl)phenol 98%

Lieferant: Apollo Scientific

Synonyme: 2'-Methyl-[1,1'-biphenyl]-4-ol

OR954765-1G OR954765-5G
APOSOR954765-1GEA 219 EUR
APOSOR954765-1G APOSOR954765-5G
4-(2-Methylphenyl)phenol 98%
4-(2-Methylphenyl)phenol
Formel: C₁₃H₁₂O
Molecular Weight: 184,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06802303
CAS-Nummer: 38262-85-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR