4-(3-Methylphenyl)phenol 96%

Lieferant: Apollo Scientific

Synonyme: 3'-Methyl-[1,1'-biphenyl]-4-ol

OR954768-1G OR954768-5G
APOSOR954768-1GEA 219 EUR
APOSOR954768-1G APOSOR954768-5G
4-(3-Methylphenyl)phenol 96%
4-(3-Methylphenyl)phenol
Formel: C₁₃H₁₂O
Molecular Weight: 184,234 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01910124
CAS-Nummer: 191724-08-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR