5-(4-Chlorophenyl)-3-pyridinecarboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR955007-1G OR955007-250MG OR955007-5G
APOSOR955007-1GEA 325 EUR
APOSOR955007-1G APOSOR955007-250MG APOSOR955007-5G
5-(4-Chlorophenyl)-3-pyridinecarboxylic acid 97%
5-(4-Chlorophenyl)-3-pyridinecarboxylic acid
Formel: C₁₂H₈ClNO₂
Molecular Weight: 233,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06410266
CAS-Nummer: 187999-33-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR