Phenylmaleic acid 95%
Lieferant: Apollo Scientific
Synonyme:
2-Phenylmaleic acid
OR955233-1G
OR955233-250MG
APOSOR955233-1GEA
352
EUR
APOSOR955233-1G
APOSOR955233-250MG
Phenylmaleic acid 95%
Phenylmaleic acid
Formel:
C₁₀H₈O₄ Molecular Weight: 192,168 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01075740 CAS-Nummer: 16110-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...