4'-Chloro-biphenyl-4-acetic acid 97%
Lieferant: Apollo Scientific
Synonyme:
2-(4'-Chloro-[1,1'-biphenyl]-4-yl)acetic acid
OR955330-1G
APOSOR955330-1GEA
149
EUR
APOSOR955330-1G
4'-Chloro-biphenyl-4-acetic acid 97%
4'-Chloro-biphenyl-4-acetic acid
Formel:
C₁₄H₁₁ClO₂ Molecular Weight: 246.689 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03426507 CAS-Nummer: 5525-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...