Methyl (E)-3-(2-bromophenyl)prop-2-enoate 96%

Lieferant: Apollo Scientific

Synonyme: Methyl (2E)-3-(2-bromophenyl)prop-2-enoate

OR955431-1G OR955431-5G
APOSOR955431-1GEA 114 EUR
APOSOR955431-1G APOSOR955431-5G
Methyl (E)-3-(2-bromophenyl)prop-2-enoate 96%
Methyl (E)-3-(2-bromophenyl)prop-2-enoate
Formel: C₁₀H₉BrO₂
Molecular Weight: 241,081 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11111009
CAS-Nummer: 92991-89-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR