Methyl 3-cyclopentylpropanoate 95%

Lieferant: Apollo Scientific

OR955455-1G OR955455-5G
APOSOR955455-1GEA 229 EUR
APOSOR955455-1G APOSOR955455-5G
Methyl 3-cyclopentylpropanoate 95%
Methyl 3-cyclopentylpropanoate
Formel: C₉H₁₆O₂
Molecular Weight: 156.222 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12031704
CAS-Nummer: 4300-02-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR