6-Phenyl-3-thio-1,2,4-triazin-5(2H)-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Phenyl-3-sulphanyl-1,2,4-triazin-5(2H)-one, 2,5-Dihydro-5-oxo-6-phenyl-3-sulphanyl-1,2,4-triazine

OR9555-1G OR9555-250MG
APOSOR9555-1GEA 143 EUR
APOSOR9555-1G APOSOR9555-250MG
6-Phenyl-3-thio-1,2,4-triazin-5(2H)-one
6-Phenyl-3-thio-1,2,4-triazin-5(2H)-one
Formel: C₉H₇N₃OS
Molecular Weight: 205,236 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00665863
CAS-Nummer: 447-00-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR