Kaffeesäuremethylester 95%

Lieferant: Apollo Scientific

Synonyme: 3,4-Dihydroxycinnamic acid methyl ester, Methyl 3,4-dihydroxycinnamate, Caffeic acid methyl ester, Methyl 3-(3,4-dihydroxyphenyl)acrylate

OR955583-1G OR955583-5G
APOSOR955583-1GEA 79.9 EUR
APOSOR955583-1G APOSOR955583-5G
Kaffeesäuremethylester 95%
Kaffeesäuremethylester
Formel: C₁₀H₁₀O₄
Molecular Weight: 194.19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00210468
CAS-Nummer: 67667-67-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR