2-Chloro-6-methyl-benzoic acid methyl ester 95%
Lieferant: Apollo Scientific
Synonyme:
Methyl 2-chloro-6-methylbenzoate
OR955603-5G
APOSOR955603-5GEA
111
EUR
APOSOR955603-5G
2-Chloro-6-methyl-benzoic acid methyl ester 95%
2-Chloro-6-methyl-benzoic acid methyl ester
Formel:
C₉H₉ClO₂ Molecular Weight: 184,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06204147 CAS-Nummer: 99585-14-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...