4-Chloro-2-methyl-benzoic acid methyl ester 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 4-chloro-2-methylbenzoate

OR955773-5G
APOSOR955773-5GEA 144 EUR
APOSOR955773-5G
4-Chloro-2-methyl-benzoic acid methyl ester 98%
4-Chloro-2-methyl-benzoic acid methyl ester
Formel: C₉H₉ClO₂
Molecular Weight: 184,62 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09842344
CAS-Nummer: 99585-12-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR