Methyl (2R)-2-aminopent-4-ynoate 97%

Lieferant: Apollo Scientific

OR956016-1G OR956016-250MG
APOSOR956016-1GEA 806 EUR
APOSOR956016-1G APOSOR956016-250MG
Methyl (2R)-2-aminopent-4-ynoate 97%
Methyl (2R)-2-aminopent-4-ynoate
Formel: C₆H₉NO₂
Molecular Weight: 127.141 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18252702
CAS-Nummer: 78342-50-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR