4-(Methanesulphonylmethyl)benzonitrile 98%
Lieferant: Apollo Scientific
Synonyme:
4-((Methylsulphonyl)methyl)benzonitrile
OR956194-1G
OR956194-5G
APOSOR956194-1GEA
179
EUR
APOSOR956194-1G
APOSOR956194-5G
4-(Methanesulphonylmethyl)benzonitrile 98%
4-(Methanesulphonylmethyl)benzonitrile
Formel:
C₉H₉NO₂S Molecular Weight: 195,238 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD15201679 CAS-Nummer: 161417-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...