2-(Methanesulphonylmethyl)benzonitrile 98%

Lieferant: Apollo Scientific

OR956198-1G OR956198-5G
APOSOR956198-1GEA 290 EUR
APOSOR956198-1G APOSOR956198-5G
2-(Methanesulphonylmethyl)benzonitrile 98%
2-(Methanesulphonylmethyl)benzonitrile
Formel: C₉H₉NO₂S
Molecular Weight: 195,238 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19257810
CAS-Nummer: 25195-58-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR