(S)-b-Amino-2-chloro-benzeneethanol 97%
Lieferant: Apollo Scientific
Synonyme:
(S)-2-Amino-2-(2-chlorophenyl)ethanol
OR956203-1G
OR956203-250MG
APOSOR956203-1GEA
1080
EUR
APOSOR956203-1G
APOSOR956203-250MG
(S)-b-Amino-2-chloro-benzeneethanol 97%
(S)-b-Amino-2-chloro-benzeneethanol
Formel:
C₈H₁₀ClNO Molecular Weight: 171,624 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09253611 CAS-Nummer: 213990-63-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...