Ethyl 2-amino-6-chlorobenzoate 95%

Lieferant: Apollo Scientific

OR956410-5G
APOSOR956410-5GEA 144 EUR
APOSOR956410-5G
Ethyl 2-amino-6-chlorobenzoate 95%
Ethyl 2-amino-6-chlorobenzoate
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08275014
CAS-Nummer: 172217-11-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR