b-Amino-2-chlorobenzenepropanol 97%
Lieferant: Apollo Scientific
Synonyme:
2-Amino-3-(2-chlorophenyl)propan-1-ol
OR956559-1G
OR956559-250MG
APOSOR956559-1GEA
314
EUR
APOSOR956559-1G
APOSOR956559-250MG
b-Amino-2-chlorobenzenepropanol 97%
b-Amino-2-chlorobenzenepropanol
Formel:
C₉H₁₂ClNO Molecular Weight: 185,651 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD10034618 CAS-Nummer: 37844-07-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...