2,3,4-Trichloropyridine 98%

Lieferant: Apollo Scientific

OR956973-1G OR956973-250MG OR956973-5G
APOSOR956973-1GEA 48 EUR
APOSOR956973-1G APOSOR956973-250MG APOSOR956973-5G
2,3,4-Trichloropyridine 98%
2,3,4-Trichloropyridine
Formel: C₅H₂Cl₃N
Molecular Weight: 182.435 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22576127
CAS-Nummer: 55934-02-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR