1-Chloro-4-(propan-2-yloxy)benzene 95%

Lieferant: Apollo Scientific

Synonyme: 1-Chloro-4-isopropoxybenzene

OR957008-1G OR957008-250MG
APOSOR957008-1GEA 487 EUR
APOSOR957008-1G APOSOR957008-250MG
1-Chloro-4-(propan-2-yloxy)benzene 95%
1-Chloro-4-(propan-2-yloxy)benzene
Formel: C₉H₁₁ClO
Molecular Weight: 170.636 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00070779
CAS-Nummer: 51241-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR