5-Chloro-2-(propan-2-yloxy)pyridine 95%

Lieferant: Apollo Scientific

Synonyme: 5-Chloro-2-isopropoxypyridine

OR957010-1G
APOSOR957010-1GEA 178 EUR
APOSOR957010-1G
5-Chloro-2-(propan-2-yloxy)pyridine 95%
5-Chloro-2-(propan-2-yloxy)pyridine
Formel: C₈H₁₀ClNO
Molecular Weight: 171,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD14705787
CAS-Nummer: 282723-23-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR