3-(N,N-Dimethylsulphamoylamino)benzeneboronic acid 96%

Lieferant: Apollo Scientific
Achtung

OR9571-1G OR9571-5G
APOSOR9571-1GEA 241 EUR
APOSOR9571-1G APOSOR9571-5G
3-(N,N-Dimethylsulphamoylamino)benzeneboronic acid 96%
3-(N,N-Dimethylsulphamoylamino)benzeneboronic acid
Formel: C₈H₁₃BN₂O₄S
Molecular Weight: 244.076 g/mol
Schmelzpunkt: 129…132 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08235107
CAS-Nummer: 277295-50-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR