6-Ethylpyridin-3-amine 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR957164-1G
OR957164-250MG
APOSOR957164-1GEA
314
EUR
APOSOR957164-1G
APOSOR957164-250MG
6-Ethylpyridin-3-amine 95%
6-Ethylpyridin-3-amine
Formel:
C₇H₁₀N₂ Molecular Weight: 122.168 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11111131 CAS-Nummer: 126553-00-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...