b-Amino-3-methoxybenzenepropanol 97%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-3-(3-methoxyphenyl)propan-1-ol

OR957211-1G OR957211-250MG OR957211-500MG
APOSOR957211-1GEA 764 EUR
APOSOR957211-1G APOSOR957211-250MG APOSOR957211-500MG
b-Amino-3-methoxybenzenepropanol 97%
b-Amino-3-methoxybenzenepropanol
Formel: C₁₀H₁₅NO₂
Molecular Weight: 181,232 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD10034616
CAS-Nummer: 344359-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR