2-Isobutoxyphenol 95%

Lieferant: Apollo Scientific

OR957528-1G OR957528-5G
APOSOR957528-1GEA 219 EUR
APOSOR957528-1G APOSOR957528-5G
2-Isobutoxyphenol 95%
2-Isobutoxyphenol
Formel: C₁₀H₁₄O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00474627
CAS-Nummer: 21315-20-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR