5-Methoxy-1,2,3,4-tetrahydroquinoline hydrochloride 95%
Lieferant: Apollo Scientific
OR958186-1G
OR958186-250MG
APOSOR958186-1GEA
139
EUR
APOSOR958186-1G
APOSOR958186-250MG
5-Methoxy-1,2,3,4-tetrahydroquinoline hydrochloride 95%
5-Methoxy-1,2,3,4-tetrahydroquinoline hydrochloride
Formel:
C₁₀H₁₃NO·ClH Molecular Weight: 199,677 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09026772 CAS-Nummer: 1073968-65-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...