4-tert-Butyl-2-chloropyrimidine 95%

Lieferant: Apollo Scientific

Synonyme: 4-(tert-Butyl)-2-chloropyrimidine

OR958233-1G OR958233-250MG
APOSOR958233-1GEA 459 EUR
APOSOR958233-1G APOSOR958233-250MG
4-tert-Butyl-2-chloropyrimidine 95%
4-tert-Butyl-2-chloropyrimidine
Formel: C₈H₁₁ClN₂
Molecular Weight: 170,639 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12402334
CAS-Nummer: 66522-06-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR