2-Cyclopropylaminonicotinic acid 95%
Lieferant: Apollo Scientific
Synonyme:
2-(Cyclopropylamino)nicotinic acid
OR958277-10G
OR958277-5G
APOSOR958277-10GEA
58
EUR
APOSOR958277-10G
APOSOR958277-5G
2-Cyclopropylaminonicotinic acid 95%
2-Cyclopropylaminonicotinic acid
Formel:
C₉H₁₀N₂O₂ Molecular Weight: 178.188 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11106728 CAS-Nummer: 639807-18-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...