5-Amino-2-phenylpyridazin-3(2H)-one 95%

Lieferant: Apollo Scientific

OR958362-10G OR958362-1G OR958362-5G
APOSOR958362-10GEA 487 EUR
APOSOR958362-10G APOSOR958362-1G APOSOR958362-5G
5-Amino-2-phenylpyridazin-3(2H)-one 95%
5-Amino-2-phenylpyridazin-3(2H)-one
Formel: C₁₀H₉N₃O
Molecular Weight: 187,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18071544
CAS-Nummer: 13589-77-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR